PocketTips™ Pipette Tips
Thermo Scientific™

PocketTips™ Pipette Tips

Eliminate time-consuming steps, reduce lab ware usage and accelerate assays with PocketTips™ Pipette Tips.
Have Questions?
Catalog NumberVolume (Metric)For Use With (Equipment)
FLIPR384B-250250 nLMolecular Devices FLIPR 384-well Head
Catalog number FLIPR384B-250
Price (USD)
-
Volume (Metric):
250 nL
For Use With (Equipment):
Molecular Devices FLIPR 384-well Head
Eliminate time-consuming pipetting steps, reduce lab ware usage and accelerate assays such as PCR primer dilutions with user-friendly PocketTips™ Pipette Tips, which are compatible on multiple workstations. The external dimensions of the tip and rack are the same as standard automation tips, requiring minimal modification and set-up. Simple, economical assay miniaturization preserves compounds and reduces reagent costs.

Features:

  • Transfer samples directly to the assay plate
  • Combine sample and assay material with soft in-tip mixing
  • Coefficient of variation (CV) of less than 6% can be achieved for any pocket size
  • Color: Clear

Recommended for:

Cell-based or low DMSO tolerant assays, selectivity assays (P450), potency determinations (IC50), compound storage and delivery from compound libraries, protein and enzyme aqueous based assays, automated clinical diagnostics and kinetic read (FLIPR) assays.

Warranty: A five (5)-year shelf life from the date of manufacture is recommended.

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
ColorClear
FilteredNon-filtered
For Use With (Equipment)Molecular Devices FLIPR 384-well Head
FormatRack
Product LinePocketTip
SterilityNon-sterile
Volume (Metric)250 nL
Unit SizeCase of 19200