Cell Lysis Buffer II
Invitrogen™

Cell Lysis Buffer II

Cell Lysis Buffer II is a high-quality, ready-to-use lysis buffer suitable for the preparation of cell extracts for ELISA, western blotting, and antibody bead immunoassays (Luminex) applications.
Have Questions?
Catalog NumberQuantity
FNN0021100 mL
Catalog number FNN0021
Price (USD)
191.65
Online exclusive
200.00
Save 8.35
Each
Quantity:
100 mL
Price (USD)
191.65
Online exclusive
200.00
Save 8.35
Each

Cell Lysis Buffer II is a high-quality, ready-to-use lysis buffer suitable for the preparation of cell extracts for ELISA, western blotting, and antibody bead immunoassays (Luminex) applications. The buffer is used for total protein extraction and utilizes detergent-based lysis, eliminating the need for mechanical cell disruption. The formulation helps retain the protein structure and function needed for enzyme assays or immunoassays.

Buffer formulation: 50 mM Tris, pH 7.4, 250 mM NaCl, 5 mM EDTA, 50 mM NaF, 1 mM Na3VO4, 1% detergent, 0.02% NaN3

This cell lysis buffer does not contain protease inhibitors and should be supplemented with 1 mM PMSF and protease inhibitor cocktail (Cat. No. A32955) just prior to use.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeCell Lysis Buffer
Quantity100 mL
Shipping ConditionDry Ice
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C

Frequently asked questions (FAQs)

What extraction reagents are recommended for efficient mouse tissue analysis?

We have 5 different cell and tissue extraction buffers suitable for preparing mouse cell and tissue extracts. These buffers can be used to extract cells and tissues from many other species as well. The exact compositions of all of our buffers are proprietary, but they are similar to those described by many researchers.

Four of these buffers can be used to prepare extracts which can be analyzed with our ELISA and Luminex kits and by Western blotting. Our Cell Extraction Buffer (FNN0011) contains extra phosphatase inhibitors and resembles the RIPA formulation that many people use. Our Tissue Extraction Reagents I (FNN0071) and II (FNN0081) contain different concentrations of NaCl and different surfactants, but are otherwise similar to each other. For those who prefer using an extraction buffer containing the detergent NP-40, we have our NP-40 Lysis Buffer (FNN0021). Finally, we sell a Denaturing Cell Extraction buffer (FNN0091) which contains 3 detergents and a chaotropic agent. Extracts prepared with FNN0091 can be analyzed with our ELISA kits and by Western blotting only. These buffers do not contain protease inhibitors, which the investigator should add right before use.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

During my ProcartaPlex assay data analysis, I am getting a warning message that there is high bead aggregation. What should I do?

Here are possible causes and solutions for this issue:

- Check the protocol settings (make sure you select the correct DD settings).
- Check the level of sheath fluid and empty the waste.
- Before acquiring the plate, run calibration and verification beads on the Luminex instrument.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

During my ProcartaPlex assay data analysis, the beads fall below or to the lower left of their bead region on the bead map. Why is this?

Here are possible causes and solutions for this issue:

This usually indicates that the beads have been photobleached. This problem can also be caused by exposing the beads to organic solvents. Unfortunately, the assay will have to be repeated because the beads cannot be restored. The beads must be protected from light and organic solvents.
Alternatively, the instrument may be off in its measurements or you may have a calibration issue. Call the manufacturer for a service appointment.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

During my ProcartaPlex assay data analysis, beads do not appear in the region gated. What happened?

This indicates that an incorrect buffer was used for the final step. The Wash Solution provided in the kit must be used for washing the beads and the Reading Buffer should be used for resuspending the beads before loading them into the Luminex instrument. The osmolarity of the solution will impact the size of the bead, and any change in the bead size will alter detection by the instrument.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

The bead counts for all of my ProcartaPlex assay wells are erratic. What went wrong?

Here are some suggestions:

- Before acquiring the plate, run calibration and verification beads on the Luminex instrument.
- Review the instrument settings and make sure they are appropriate for the assay being run (adjustment of needle height, make sure you select the correct bead gates and the correct DD settings).
- Shake the plate before acquisition on the instrument to resuspend the beads.
- Vortex the beads for 30 sec before adding them into the plate.
- Washing: Do not forget to keep the plate for about 2 mins on the Hand-Held Magnetic Plate Washer before emptying the plate.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Citations & References (8)

Citations & References
Abstract
Cleavage of hyaluronan is impaired in aged dermal wounds.
Authors:Reed MJ, Damodarasamy M, Chan CK, Johnson MN, Wight TN, Vernon RB,
Journal:Matrix Biol
PubMed ID:23022999
'Changes in extracellular matrix (ECM) are one of many components that contribute to impaired wound healing in aging. This study examined the effect of age on the glycosaminoglycan hyaluronan (HA) in normal and wounded dermis from young (4-6 month-old) and aged (22-24 month-old) mice. HA content and size were similar ... More
Thyroid hormone controls the gene expression of HSV-1 LAT and ICP0 in neuronal cells.
Authors:Bedadala GR, Pinnoji RC, Palem JR, Hsia SC,
Journal:Cell Res
PubMed ID:20386570
'Various factors/pathways including hormonal regulation have been suggested to control herpes simplex virus type 1 (HSV-1) latency and reactivation. Our computer analysis identified a DNA repeat containing thyroid hormone-responsive elements (TRE) in the regulatory region of HSV-1 latency-associated transcript (LAT). Thyroid hormone (triiodothyronine, T(3)) functions via its receptor TR (thyroid ... More
miRNA-214 is related to invasiveness of human non-small cell lung cancer and directly regulates alpha protein kinase 2 expression.
Authors:Salim H, Arvanitis A, de Petris L, Kanter L, Hååg P, Zovko A, Özata DM, Lui WO, Lundholm L, Zhivotovsky B, Lewensohn R, Viktorsson K,
Journal:
PubMed ID:23929716
'The prognosis of non-small cell lung cancer (NSCLC) is poor, since it has often metastasized to distant organs by the time of diagnosis. Therefore, biomarkers predicting metastasis are crucial. miRNAs play important roles in the regulation of different tumor cell processes, including metastasis. We recently showed that miRNA-214 is linked ... More
miR-205 expression promotes cell proliferation and migration of human cervical cancer cells.
Authors:Xie H, Zhao Y, Caramuta S, Larsson C, Lui WO,
Journal:PLoS One
PubMed ID:23056551
MicroRNAs (miRNAs) are short non-coding RNA regulators that control gene expression mainly through post-transcriptional silencing. We previously identified miR-205 in a signature for human cervical cancer using a deep sequencing approach. In this study, we confirmed that miR-205 expression was frequently higher in human cervical cancer than their matched normal ... More
A preclinical assessment of neural stem cells as delivery vehicles for anti-amyloid therapeutics.
Authors:Njie eG, Kantorovich S, Astary GW, Green C, Zheng T, Semple-Rowland SL, Steindler DA, Sarntinoranont M, Streit WJ, Borchelt DR,
Journal:PLoS One
PubMed ID:22496779
Transplantation of neural stems cells (NSCs) could be a useful means to deliver biologic therapeutics for late-stage Alzheimer's disease (AD). In this study, we conducted a small preclinical investigation of whether NSCs could be modified to express metalloproteinase 9 (MMP9), a secreted protease reported to degrade aggregated Aß peptides that ... More