Product Image
Invitrogen™

Denaturing Cell Extraction Buffer

変性細胞抽出バッファーはELISAやウェスタンブロッティングでの使用に適しています。これは、サンプルの処理または沸騰が必要なInvitrogen phospho-ELISA(ERK、JNK)に使用できます。変性細胞抽出バッファーで細胞を溶解した後、溶解物でのサンプル処理や沸騰は必要ありません。用途:サンプル⁄アッセイの調製詳細を見る
製品番号(カタログ番号)数量
FNN0091100 ml
製品番号(カタログ番号) FNN0091
価格(JPY)
34,000
Each
数量:
100 ml
変性細胞抽出バッファーはELISAやウェスタンブロッティングでの使用に適しています。これは、サンプルの処理または沸騰が必要なInvitrogen phospho-ELISA(ERK、JNK)に使用できます。変性細胞抽出バッファーで細胞を溶解した後、溶解物でのサンプル処理や沸騰は必要ありません。

用途:サンプル⁄アッセイの調製
研究用にのみ使用できます。診断用には使用いただけません。
仕様
製品タイプ細胞抽出バッファー
数量100 ml
出荷条件ドライアイス
Unit SizeEach
組成および保存条件
Store at 2°C to 8°C

よくあるご質問(FAQ)

What extraction reagents are recommended for efficient mouse tissue analysis?

We have 5 different cell and tissue extraction buffers suitable for preparing mouse cell and tissue extracts. These buffers can be used to extract cells and tissues from many other species as well. The exact compositions of all of our buffers are proprietary, but they are similar to those described by many researchers.

Four of these buffers can be used to prepare extracts which can be analyzed with our ELISA and Luminex kits and by Western blotting. Our Cell Extraction Buffer (FNN0011) contains extra phosphatase inhibitors and resembles the RIPA formulation that many people use. Our Tissue Extraction Reagents I (FNN0071) and II (FNN0081) contain different concentrations of NaCl and different surfactants, but are otherwise similar to each other. For those who prefer using an extraction buffer containing the detergent NP-40, we have our NP-40 Lysis Buffer (FNN0021). Finally, we sell a Denaturing Cell Extraction buffer (FNN0091) which contains 3 detergents and a chaotropic agent. Extracts prepared with FNN0091 can be analyzed with our ELISA kits and by Western blotting only. These buffers do not contain protease inhibitors, which the investigator should add right before use.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

During my ProcartaPlex assay data analysis, I am getting a warning message that there is high bead aggregation. What should I do?

Here are possible causes and solutions for this issue:

- Check the protocol settings (make sure you select the correct DD settings).
- Check the level of sheath fluid and empty the waste.
- Before acquiring the plate, run calibration and verification beads on the Luminex instrument.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

During my ProcartaPlex assay data analysis, the beads fall below or to the lower left of their bead region on the bead map. Why is this?

Here are possible causes and solutions for this issue:

This usually indicates that the beads have been photobleached. This problem can also be caused by exposing the beads to organic solvents. Unfortunately, the assay will have to be repeated because the beads cannot be restored. The beads must be protected from light and organic solvents.
Alternatively, the instrument may be off in its measurements or you may have a calibration issue. Call the manufacturer for a service appointment.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

During my ProcartaPlex assay data analysis, beads do not appear in the region gated. What happened?

This indicates that an incorrect buffer was used for the final step. The Wash Solution provided in the kit must be used for washing the beads and the Reading Buffer should be used for resuspending the beads before loading them into the Luminex instrument. The osmolarity of the solution will impact the size of the bead, and any change in the bead size will alter detection by the instrument.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

The bead counts for all of my ProcartaPlex assay wells are erratic. What went wrong?

Here are some suggestions:

- Before acquiring the plate, run calibration and verification beads on the Luminex instrument.
- Review the instrument settings and make sure they are appropriate for the assay being run (adjustment of needle height, make sure you select the correct bead gates and the correct DD settings).
- Shake the plate before acquisition on the instrument to resuspend the beads.
- Vortex the beads for 30 sec before adding them into the plate.
- Washing: Do not forget to keep the plate for about 2 mins on the Hand-Held Magnetic Plate Washer before emptying the plate.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

引用および参考文献 (4)

引用および参考文献
Abstract
The PAXgene(®) tissue system preserves phosphoproteins in human tissue specimens and enables comprehensive protein biomarker research.
Authors:Gündisch S, Schott C, Wolff C, Tran K, Beese C, Viertler C, Zatloukal K, Becker KF,
Journal:PLoS One
PubMed ID:23555997
'Precise quantitation of protein biomarkers in clinical tissue specimens is a prerequisite for accurate and effective diagnosis, prognosis, and personalized medicine. Although progress is being made, protein analysis from formalin-fixed and paraffin-embedded tissues is still challenging. In previous reports, we showed that the novel formalin-free tissue preservation technology, the PAXgene ... More
CaMKII and MEK1/2 inhibition time-dependently modify inflammatory signaling in rat cerebral arteries during organ culture.
Authors:Waldsee R, Eftekhari S, Ahnstedt H, Johnson LE, Edvinsson L,
Journal:
PubMed ID:24886705
'Cerebral ischemia induces transcriptional upregulation of inflammatory genes in the brain parenchyma and in cerebral arteries, thereby contributing to the infarct development. The present study was designed to evaluate the involvement of calcium-calmodulin-dependent protein kinase (CaMKII) II and extracellular signal-regulated kinase1/2 (ERK1/2) on inflammatory mediators in rat cerebral arteries using ... More
Altered MARCH1 ubiquination-regulated dendritic cell immune functions during the early stage of zymosan-induced multiple organ dysfunction syndrome (MODS) in mice.
Authors:Li F, Lu JY, Liu Q, Wang HW, Guo H,
Journal:Immunol Lett
PubMed ID:23305794
Using a zymosan-induced mouse model of multiple organ dysfunction syndrome (MODS), we previously found profound increases in spleen immune cells' expressions of ubiquitin and MHC-II molecules and increased CD11c+ dendritic cells (DCs) within 24h of zymosan injection. We postulated that the early stage of MODS altered DCs function via an ... More
Enhancement of cisplatin cytotoxicity by benzyl isothiocyanate in HL-60 cells.
Authors:Lee Y, Kim YJ, Choi YJ, Lee JW, Lee S, Chung HW,
Journal:Food Chem Toxicol
PubMed ID:22525867
Cis-diamminedichloroplatinum (II) (cisplatin) is one of the most widely used chemotherapeutic drugs, but its effectiveness is limited by tumor cell resistance and the severe side effects it causes. One strategy for overcoming this problem is the concomitant use of natural dietary compounds as therapeutic agents. Benzyl isothiocyanate (BITC) is a ... More