E-Gel™ EX Agarose Gels, 1%, 20 Gels/Pk. - FAQs
E-Gel™ EX Agarose Gels, 1%, 20 Gels/Pk. - FAQs
View additional product information for E-Gel™ EX Agarose Gels - FAQs (G401001, G402021, G402022, G401002, G401004)
2 product FAQs found
What ladder do you recommend for running RNA on Invitrogen E-Gel EX Agarose Gels?
Depending on the RNA fragment size, we recommend the following ladders:
  • RiboRuler Low Range RNA ladder, (100 bp-1,000 bp) (Cat. No. SM1833)
  • RiboRuler High Range RNA ladder, (200 bp-6,000 bp) (Cat. No. SM1823)
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Gel Electrophoresis and Blotting Support Center.
Can you provide me with the protocol to run E-Gel EX gels for RNA samples on the E-Gel PowerSnap Electrophoresis System?
Yes, the protocol for running E-Gel EX gels for RNA samples on the E-Gel PowerSnap Electrophoresis System can be found in the E-Gel Power Snap Electrophoresis System User Guide, on page 46 under Appendix E.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Gel Electrophoresis and Blotting Support Center.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Gel Electrophoresis and Blotting Support Center.