E-Gel™ Agarose Gels with SYBR Safe™ DNA Gel Stain Starter Kit, 1.2% - FAQs
E-Gel™ Agarose Gels with SYBR Safe™ DNA Gel Stain Starter Kit, 1.2% - FAQs
View additional product information for E-Gel™ Agarose Gels with SYBR Safe™ DNA Gel Stain Starter Kit, 1.2% - FAQs (G620601)
1 product FAQs found
What is the function of the E-Editor 2.0 software?
The E-Editor 2.0 software allows you to quickly reconfigure digital images of E-Gel 48, E-Gel 96, E-PAGE 48 and E-PAGE 96 gel results for analysis and documentation. You can capture an image of the gel and then use the E-Editor 2.0 software to:
-Align and arrange the lanes in the image to any 48, 96, or 384 image
-Save the reconfigured image for further analysis
-Copy and paste selected lanes or the entire image into other applications for printing, saving, e-mailing, and/or publishing on the Web.
The E-Editor 2.0 does not take perform densitometry analysis from your gel images. The E-Editor 2.0 can be downloaded for free.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.