E-Gel™ iBase™ and E-Gel™ Safe Imager™ Combo Kit (UK only) - FAQs
E-Gel™ iBase™ and E-Gel™ Safe Imager™ Combo Kit (UK only) - FAQs
View additional product information for E-Gel™ iBase™ and E-Gel™ Safe Imager™ Combo Kit (UK only) - FAQs (G6465UK)
2 product FAQs found
My E-Gel iBase/PowerBase/E-Base system is continuously beeping and/or shows a steady red light. What am I doing wrong?
Check the E-Gel cassette and copper contacts. You can try replacing the gel cassette with a fresh gel cassette to see whether this fixes the problem. A cold cassette or improper operating conditions can also lead to this failure. Cassettes should be at room temperature for use; avoid storing at 4°C.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.
What is the difference between the E-Gel Safe Imager Real-Time Transilluminator (Cat. No. G6500) and the Safe Imager 2.0 Blue-Light Transilluminator (Cat. No. G6600)?
The E-Gel Safe Imager Real-Time Transilluminator docks onto the E-Gel iBase Power System and allows real-time imaging of the migration of DNA/RNA in E-Gel Agarose Gels containing SYBR Safe stain. The Safe Imager 2.0 Blue-Light Transilluminator is an independent system designed for viewing stained gels on the benchtop.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.