E-Gel™ 96 Agarose Gels, 2% with E-Gel™ Low Range Ladder - FAQs
E-Gel™ 96 Agarose Gels, 2% with E-Gel™ Low Range Ladder - FAQs
View additional product information for E-Gel™ 96 Agarose Gels, 2% with E-Gel™ Low Range Ladder - FAQs (G700811)
1 product FAQs found
Are there robotic scripts for running E-Gel 48/96 or E-PAGE 48/96 gels?
Robotic scripts for running E-Gel 96 Agarose Gels and E-PAGE 96 Gels on Beckman Coulter's Biomek FX workstation can be found on our website at: www.thermofisher.com/egels (click the Labware Definitions link in the left navigation pane).
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.