E-Gel™ Power Snap Electrophoresis Device Starter Kit, CloneWell II 0.8% - FAQs
E-Gel™ Power Snap Electrophoresis Device Starter Kit, CloneWell II 0.8% - FAQs
View additional product information for E-Gel™ Power Snap Electrophoresis Device Starter Kit, CloneWell II 0.8% - FAQs (G8168ST)
1 product FAQs found
I accidentally allowed my DNA to run past the collection wells for my E-Gel SizeSelect II or E-Gel CloneWell II agarose gels. What should I do?
You can use the Reverse E-Gel Program to run the band back into the collection gel.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.