E-Gel™ 48 Agarose Gels
Invitrogen™

E-Gel™ 48 Agarose Gels

Invitrogen E-Gel 48 high-throughput precast agarose gels with or without SYBR Safe DNA Gel Stain are designed for fast, convenient, and less-hazardous DNA sample electrophoresis.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량분리 범위젤 비율dimStainType
G8208424 x 8 Gels50 to 3000 bp2%
G8008018 Gels400 to 10,000 bp1%
G8208018 Gels400 to 10,000 bp1%
G8208414 x 8 Gels400 to 10,000 bp1%
G8008028 Gels50 to 3000 bp2%
G8208028 Gels50 to 3000 bp2%
G8008048 Gels10 to 400 bp4%
G8208048 Gels10 to 500 bp4%
G8208444 x 8 Gels10 to 500 bp4%
카탈로그 번호 G820842
제품 가격(KRW)
1,308,000
온라인 행사
Ends: 30-Sep-2026
1,453,000
할인액 145,000 (10%)
Each
수량:
4 x 8 Gels
분리 범위:
50 to 3000 bp
젤 비율:
2%
제품 가격(KRW)
1,308,000
온라인 행사
Ends: 30-Sep-2026
1,453,000
할인액 145,000 (10%)
Each
Invitrogen E-Gel 48 high-throughput precast agarose gels with or without SYBR Safe DNA Gel Stain are designed for fast, convenient, and less-hazardous DNA sample electrophoresis. Each E-Gel agarose gel cassette contains all components required for efficient gel separation and analysis—just load your samples and run.

Each gel contains 48 sample wells and 4 marker wells in 1%, 2%, or 4% high-resolution agarose with a 3.2-cm run length. The various agarose percentages are used to separate DNA fragments of different sizes:

  • 1% agarose gels: DNA fragments between 400 bp and 10 kb
  • 2% agarose gels: DNA fragments between 50 bp and 3 kb
  • 4% agarose gels: DNA fragments between 10 bp and 500 bp

Each cassette is individually labeled with an EAN39 bar code that is recognized by most commercially available robotic bar code readers.

Features of E-Gel gels:

  • High-throughput analysis—contains 48 sample lanes and 4 marker lanes per gel
  • Automation compatible—suitable for use with multi-channel pipettors and commonly used 8- and 12-pin liquid handling robots
  • Scalable—resolve up to 192 samples in one run by combining up to four E-Base Mother and E-Base Daughter device units
  • Less hazardous choice—contains SYBR Safe DNA stain, a less hazardous alternative to ethidium bromide
  • High performance—detect 3 ng of sample DNA with separation time as little as ∼20 minutes

Less hazardous option

Choose gels with SYBR Safe DNA stain to minimize exposure to hazardous and mutagenic factors such as ethidium bromide and UV. Precast E-Gel agarose gels with SYBR Safe stain, combined with a blue-light trans-illumination, offer better sample integrity and a less hazardous work environment. Unlike ethidium bromide, the DNA stain most commonly incorporated into agarose gels, SYBR Safe DNA Gel Stain is a non-toxic, non-mutagenic DNA stain that is less hazardous for the user, less hazardous for the environment, and can save institutional overhead costs for disposal.

Fast and convenient analysis

E-Gel agarose gels enable fast and high-performance analysis with a detection sensitivity of 3 ng sample DNA per band and a separation time three times faster than that of conventional pour-your-own gels. Each E-Gel agarose gel contains agarose, electrodes, SYBR Safe DNA stain, and a bufferless ion exchange system packaged inside a dry disposable cassette. E-Gel technology does not require any gel or buffer preparation or gel staining steps. Just load your samples and run.

Running and imaging devices

High-throughput E-Gel agarose gels require the Invitrogen E-Base Electrophoresis Device for gel running.

Available in a variety of gel formats

E-Gel agarose gels are available in variety of gel percentage, stain, and well formats. The high-throughput E-Gel 96 and 48 agarose gels with SYBR Safe stain are available in 1%, 2%, and 4% gel percentages.

For Research Use Only. Not for use in diagnostic procedures.
사양
Compatible LaddersE-Gel™ 1 Kb Plus Express DNA Ladder
수량4 x 8 Gels
배송 조건Room Temperature
젤 비율2%
젤 유형E-Gel
라벨 또는 염료SYBR Safe
제품 유형Agarose Gel
분리 범위50 to 3000 bp
48 + 4-well
Unit SizeEach
구성 및 보관
• Each box contains 8 gels

Store strictly at room temperature. Do not freeze.
Avoid cold spots near freezers or other uncontrolled environments.

자주 묻는 질문(FAQ)

How can I view DNA on my E-Gel agarose gel with SYBR Safe DNA stain?

SYBR Safe-stained gels can be visualized with a standard UV transilluminator, a laser-based scanner equipped with an excitation source in the UV range or between 470 and 530 nm, or a blue-light transilluminator.

Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.

My E-Gel agarose gel has speckles when viewed in the imager. What should I do?

Here are some suggestions:

- Try cleaning the cassettes with alcohol and Kimwipes wipers.
- Try cleaning the camera lens.
- Try to adjust the exposure time and brightness options of the documentation system you are using.

Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.

My sample is leaking from the wells when running my E-Gel agarose gels. What happened?

Please ensure that you have not overloaded the well and that the wells were not damaged during comb removal.

Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.

I accidentally stored my E-Gel agarose gels at 4 degrees C instead of room temperature. Can I still use them?

While we recommend storage at room temperature, these gels will still be usable. Bring the gels to room temperature prior to the run for optimal conditions.

Find additional tips, troubleshooting help, and resources within ourNucleic Acid Gel Electrophoresis and Blotting Support Center.

What loading buffer should I use for my E-Gel agarose gels?

Loading buffer is optional. Samples can be loaded directly into the wells if no buffer is used, or you can dilute them with deionized water or TE buffer. If you want to use a loading buffer, please see the recipes below:

E-Gel agarose gels (including EX)
10 mM Tris-HCl, pH 7.5
1 mM EDTA
0.005% bromophenol blue
0.005% xylene cyanol FF
E-Gel CloneWell II and E-Gel SizeSelect II agarose gels
10 mM Tris-HCl, pH 7.5
1 mM EDTA

Alternatively, you can use 10X BlueJuice Gel Loading Buffer or TrackIt Loading Buffer. Dilute this buffer 50- to 200-fold to obtain optimal results with E-Gel agarose gels.

Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.