E-Gel™ Power Snap Plus Electrophoresis Systems, Accessories with Extended Warranty
E-Gel™ Power Snap Plus Electrophoresis Systems, Accessories with Extended Warranty
E-Gel™ Power Snap Plus Electrophoresis Systems, Accessories with Extended Warranty
Invitrogen™
E-Gel™ Power Snap Plus Electrophoresis Systems, Accessories with Extended Warranty
The all-in-one Invitrogen™ E-Gel™ Power Snap Plus Electrophoresis System is designed for fast and convenient high-throughput E-Gel agarose gel electrophoresis, from DNA sample loading to gel image.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeIncludesGel Type
G9201CameraCamera onlyNot Included
G9101Horizontal Agarose Electrophoresis UnitElectrophoresis device onlyNot Included
G9301Electrophoresis System Starter KitCamera, WiFi DongleE-Gel, E-PAGE
G9341Electrophoresis System Starter KitCamera, Gel, WiFi DongleE-Gel 48 1% with SYBR Safe
G9342Electrophoresis System Starter KitCamera, Gel, WiFi DongleE-Gel 48 2% with SYBR Safe
G9391Electrophoresis System Starter KitCamera, Gel, WiFi DongleE-Gel 96 1% with SYBR Safe
G9392Electrophoresis System Starter KitCamera, Gel, WiFi DongleE-Gel 96 2% with SYBR Safe
Catalog number G9201
Price (USD)
3,630.12
Each
Type:
Camera
Includes:
Camera only
Gel Type:
Not Included
Price (USD)
3,630.12
Each
The all-in-one Invitrogen E-Gel Power Snap Plus Electrophoresis System is designed for fast and convenient high-throughput E-Gel agarose gel electrophoresis, from DNA sample loading to gel imaging. With dry precast E-Gel agarose gel technology, you can run DNA samples in as little as 10 minutes and observe sample separation in real time. It is the only benchtop system that seamlessly integrates high-throughput DNA sample separation and gel imaging into one workflow.
Features of the E-Gel Power Snap Plus Electrophoresis system include:
• Streamline electrophoresis and image capture with an all-in-one system
• Compatible with 11- to 96-well E-Gel agarose gels
• View and capture DNA samples in real-time
• Access your images anywhere using the built-in Thermo Fisher Connect and network compatibility
Small, fast, and powerful
The all-in-one E-Gel Power Snap Plus Electrophoresis System provides everything you need for DNA gel electrophoresis and gel visualization in a small lightweight benchtop system. The system includes an electrophoresis power supply, blue-light transilluminator, amber filter, and adapter for 11- to 22-well E-Gel agarose gels. This enables gel separation and real-time sample tracking on E-Gel agarose gels pre-stained with SYBR Safe or SYBR Gold II DNA stains. The device arrives with pre-programmed protocols for each type of compatible E-Gel agarose gel.
Instant capture and seamless integration
Capture high-resolution E-Gel agarose gel images with the included easy-to-use E-Gel Power Snap Plus Camera. The camera is directly docked to the E-Gel Power Snap Electrophoresis Device, requiring no external power supply or connection to a desktop computer. The E-Gel Power Snap Plus Camera is also compatible with the included WiFi dongle and has an ethernet port, so you can export your experimental data to the cloud or internal servers for maximum convenience.
Unsurpassed convenience of dry precast E-Gel agarose gel
Run DNA samples in as little as 10 minutes with dry, precast, pre-stained E-Gel agarose gels. The E-Gel dry precast agarose gel technology provides speed and convenience, eliminating any need for gel preparation or staining. E-Gel agarose gels are available in a variety of gel percentage and a choice of two DNA stains. The E-Gel Power Snap Plus Electrophoresis Device is compatible with high-throughput E-Gel 48 and E-Gel 96 gels with SYBR Safe*. A provided adapter extends compatibility to low-throughput E-Gel precast agarose gels, including E-Gel EX, E-Gel SYBR Safe, E-Gel CloneWell II, and E-Gel SizeSelect II gels*.
Safer workflows
The blue light emitted by the E-Gel Power Snap Plus Electrophoresis Device transilluminator ensures maximum possible safety to the user and the samples. A combination of blue light and amber filter also offers real-time viewing of the migration of your samples. The E-Gel Power Snap Plus system is a perfect choice for routine PCR fragment analysis, genotyping, and other applications requiring high-throughput. The compatibility with E-Gel CloneWell II gels also makes it a great choice for a safer cloning workflow with increased efficiency.
Made for convenience
The integrated E-Gel Power Snap Plus Camera (available separately as G9201) touch-screen and on-board software provide an intuitive interface for quick gel image capture and editing. The device also offers built in E-Gel 96 deconvolution into several more readable formats. You can store images in the internal memory of the Power Snap Plus system, on our Thermo Fisher Connect cloud-based platform, or on local network drive to further optimize your workflow.
REX Extended Warranty
Keep your electrophoresis system up and running with an extended warranty that covers all costs for shipping, parts, and labor for repairs.
*The E-Gel Power Snap Plus Electrophoresis System also supports ethidium bromide-stained E-Gel high- and low- throughput agarose gels. The ethidium bromide-stained gels are not suitable for imaging with the E-Gel Power Snap Camera or real-time view.
For Research Use Only.
Specifications
ColorWhite
Gel TypeNot Included
IncludesCamera only
Quantity1 Camera
SterilityNon-sterile
Warranty1-year rapid exchange extended warranty included
For Use With (Application)E-Gel™ Agarose Gel Electrophoresis
For Use With (Equipment)E-Gel™ Power Snap Plus Electrophoresis Device
TypeCamera
Unit SizeEach
Frequently asked questions (FAQs)
Do you offer the adapter or 11-22 well E-Gels with the E-Gel Power Snap Plus Electrophoresis System?
Yes, the adaptor is stored in a drawer in the instrument itself. It extends the compatibility to 11-22 well E-Gels as well as E-Gel CloneWell II Agarose Gels with SYBR Safe, 0.8% (Cat. No. G661818), SizeSelect II, and E-Gel NGS.
Can I connect my E-Gel Power Snap Plus Electrophoresis System to WiFi without the WiFi dongle?
No, the E-Gel Power Snap Electrophoresis System requires the WiFi dongle to connect to the internet. The dongle is included with the Cat. No. G9301 system and all starter kits.
In the E-Gel Power Snap Plus Electrophoresis System, the matt surface of the transilluminator does not look even. Is this a defect?
No, this is expected from the special treatment of the glass. It will have no effect on the final image.
Is the E-Gel Power Snap Plus Electrophoresis System compatible with E-Gel Go! gels?
The E-Gel Power Snap Plus Electrophoresis System is not compatible with E-Gel Go! Gels.
With the E-Gel Power Snap Plus Electrophoresis System, can the E-gel well adapter be easily taken out and put back in?
Yes. There is plenty of space for fingers to grasp the E-gel well adapter in the instrument for best operation.