E-Gel™ Power Snap Electrophoresis System Starter Package, 48-well, 1%, 1-year warranty, 1 kit - FAQs
E-Gel™ Power Snap Electrophoresis System Starter Package, 48-well, 1%, 1-year warranty, 1 kit - FAQs
View additional product information for E-Gel™ Power Snap Plus Electrophoresis Systems with 1-year Standard Warranty - FAQs (G9311, G9200, G9332, G9382, G9381, G9331, G9110)
1 product FAQs found
With the E-Gel Power Snap Plus Electrophoresis System, the camera failed to startup on the first time of use. What should I do?
Try to re-dock the instrument one more time. This should fix the initial error.