Lysing Solution, Whole Blood Lysing Solution, 25 mL - FAQs
Lysing Solution, Whole Blood Lysing Solution, 25 mL - FAQs
View additional product information for Lysing Solution, Whole Blood Lysing Solution - FAQs (GAS010, GAS010S100)
1 product FAQs found
What reagents can I use to lyse red blood cells other than the FIX & PERM Cell Permeabilization Kit?
Cal-Lyse premixed lysis solution (Cat. Nos. GAS010 and GAS010S100) is specifically formulated to lyse erythrocytes following monoclonal antibody staining of whole blood or bone marrow. Treatment with this reagent leads to both the lysis of red blood cells and the fixation of white cells. Treatment does not affect fluorophore-labeled antibodies bound to leukocytes, and leaves morphological scatter characteristics of leukocytes intact.
-High-Yield Lyse (Cat. No. HYL250) is a premixed, fixative-free erythrocyte-lysis solution that can be used to help eliminate red cells from whole blood for flow cytometric analysis with minimal loss of rare blood cell populations. Using this reagent, lysis of erythrocytes is performed immediately following staining of the blood samples with fluorophore-conjugated monoclonal antibodies; there is no need for a wash step. Both are IVD reagents.