HyperSep™ PFAS Cartridges
Thermo Scientific™

HyperSep™ PFAS Cartridges

HyperSep SPE PFAS SPE products are designed to address the needs of extraction and concentration of PFAS samples for targeted and not targeted screening while minimizing background contamination.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberBed WeightParticle SizeSorbent
GCB-1010 g55 to 65 μm and 120/400 meshGraphitized Carbon Black (GCB)
60107-30-3-WAX30 mg55 to 65 μmPolymeric WAX
Catalog number GCB-10
Price (USD)
284.04
Pack of 30
Bed Weight:
10 g
Particle Size:
55 to 65 μm and 120/400 mesh
Sorbent:
Graphitized Carbon Black (GCB)
Price (USD)
284.04
Pack of 30
HyperSep SPE PFAS SPE products are designed to address the needs of extraction and concentration of PFAS samples for targeted and not targeted screening while minimizing background contamination. The range of selectivity’s and formats are designed to meet the requirements for main environmental testing methods by extracting and concentrating samples from drinking water, ground water, surface water, wastewater, soil, food and other complex matrices.

Per- and polyfluorinated alkyl substances (PFAS) are among the primary emerging contaminants of concern. The detection and quantification of known PFAS and the discovery of unknown PFAS substances is rapidly growing. SPE solutions are a fundamental part of the analytical workflow for analysis of PFAS compounds.

Thermo Scientific provides a range of products to address PFAS testing requirements e.g. EPA methods: 1633, 537.1, 533.

Specifications
DescriptionHyperSep LOOSE CARBON
For Use With (Application)EPA method 1633, extraction of PFAS from water, tissue, soil, or biosolids in conjunction with SPE cartridge
Quantity10 g
Bed Weight10 g
For Use With (Equipment)Thermo Scientific HyperSep-96 System
Particle Size55 to 65 μm and 120/400 mesh
Product LineHyperSep
SorbentGraphitized Carbon Black (GCB)
TypeSolid Phase Extraction Column
Unit SizePack of 30