Orion™ Combination pH Electrodes
Orion™ Combination pH Electrodes
Orion™ Combination pH Electrodes
Orion™ Combination pH Electrodes
Orion™ Combination pH Electrodes
Thermo Scientific™

Orion™ Combination pH Electrodes

Environmentally friendly lab pH electrodes to meet RoHS requirements by containing no lead, mercury or other hazardous substances — making disposal of old electrodes easy and hassle free.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFilling Solution TypeJunction
GD9156BNWPLiquid-filledDouble
GD9106BNWPGel-filledDouble
GS9106BNWPGel-filledSingle
Catalog number GD9156BNWP
Price (USD)
257.40
Online exclusive
286.00
Save 28.60
Each
Filling Solution Type:
Liquid-filled
Junction:
Double
Price (USD)
257.40
Online exclusive
286.00
Save 28.60
Each
Thermo Scientific™ Orion™ Green Electrodes are the first lab electrode to contain no lead, mercury or other hazardous substances and are completely ROHS compliant. GD9156BNWP is a refillable electrode that provides reliable and accurate measurements of routine samples but are best suited for treated effluent, distilled water and wastewater. GD9106BNWP and GS9106BNWP contain a low-maintenance gel that is best suited for field or plant use and educational institutes, where minimal maintenance is required.
 

Orion™ Double Junction Electrodes have an isolated Ag/AgCl reference system which prevents silver from coming in contact with the sample. Great for measuring TRIS buffer, sulfide and protein samples. While Orion™ Single Junction Electrodes provide a reliable and accurate option for routine pH measurement of typical samples.

Solutions required:

Electrode fill solution for GD9156BNWP

  • 910008 3M KCl filling solution, 5 x 60 mL bottles

Electrode storage solution

  • 910001 pH electrode storage solution, 475 mL bottle

pH buffers

  • 910168 pH 1.68 buffer, NIST traceable, 475 mL
  • 910104 pH 4.01 buffer, color coded red, NIST traceable, 475 mL
  • 910105 pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
  • 910686 pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
  • 910107 pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL
  • 910918 pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
  • 910110 pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
  • 910112 pH 12.46 buffer, NIST traceable, 475 mL
  • 910199 Standard All-in-One pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475 mL each, pH electrode storage solution, 475ml, and 12 mm electrode storage bottle

pH cleaning solution kit

  • 900020 Cleaning solution A and C, 30 mL each. B and D, 60 mL each

Items included with GD9156BNWP

  • 910008 3M KCl filling solution 60 mL 1 bottle

Warranty period

  • GD9156BNWP: 12 months from date of purchase
  • GD9106BNWP and GS9106BNWP: 6 months from date of purchase
Specifications
DescriptionLiquid filled
Filling Solution TypeLiquid-filled
For Use With (Equipment)Standard pH Meter
Length (English) Cable39.3 in.
Length (Metric) Cable1 m
Connector TypesBNC (Waterproof)
JunctionDouble
MaterialEpoxy Body
Temperature (Metric) Operating0°C to 90°C
TypeGreen Combination pH Electrode, liquid-filled
pH Range0 to 14
Unit SizeEach