Monkey MRP2 Vesicles - FAQs
Monkey MRP2 Vesicles - FAQs
View additional product information for Monkey MRP2 Vesicles - FAQs (GM0018)
3 product FAQs found
What is the supplied quantity of ATP-binding cassette (ABC) Transporter Vesicles?
The quantity supplied is 2.5 mg total protein/500 µL per vial. The protein concentration is 5 mg/mL. When our recommended protocol is followed, 50 vesicular transport assays or 125 ATPase assays can be performed with one vial of ABC Transporter Vesicles product.
Find additional tips, troubleshooting help, and resources within our Drug Discovery & Development Support Center.
What is the storage condition for ATP-binding cassette (ABC) Transporter Vesicles?
We recommend storing them at -80 degrees C.
Find additional tips, troubleshooting help, and resources within our Drug Discovery & Development Support Center.
What are ATP-binding cassette (ABC) Transporter Vesicles?
Prepared from Sf9 cells which have been engineered to over-express specific ABC transporters, ATP-binding cassette (ABC) Transporter Vesicles are “inside-out” vesicles designed for use in vesicular transport assays. They allow the investigation of possible interactions between specific ABC transporters and test compounds. ABC Transporter Vesicles provide high levels of transporter activity with low background, giving a clear signal if the test compound is a substrate or inhibitor of a specific efflux transporter.
While ABC transporters typically mediate the efflux of substrates from cells, transporters expressed on these inside-out vesicles import substrates into the vesicles. It is therefore possible to quantitatively evaluate transport activity for the test compound by determining the amount incorporated into the vesicles.
Note: One vial of vesicle product can perform 50 vesicular transport assays.
Find additional tips, troubleshooting help, and resources within our Drug Discovery & Development Support Center.