Sensititre™ GPALL1F
Sensititre™ GPALL1F
Sensititre™ GPALL1F
Thermo Scientific™

Sensititre™ GPALL1F

Perform accurate AST of non-fastidious Gram-positive isolates on a single platform with this convenient plate, featuring D-Test and Cefoxitin Screen.
Have Questions?
Catalog NumberQuantity
GPALL1F10/Pk.
Catalog number GPALL1F
Price (USD)
-
Quantity:
10/Pk.

Confidently perform susceptibility testing of non-fastidious Gram-positive isolates with Thermo Scientific™ Sensititre™ Gram Positive GPALL1F AST Plate, featuring D-Test and Cefoxitin Screen.

Test for non-fastidious Gram-positive isolates with true MIC results using Sensititre Gram Positive GPALL1F AST Plate, featuring D-Test and Cefoxitin Screen. 

  • The plate has fewer limitations for Enterococcus spp. and Staphylococcus spp. versus other automated systems
  • Full-range, true MIC readings provide results for better detection of resistance
  • Automated or manual read options are available for ultimate flexibility
  • 18 - 24-month shelf life and room-temperature storage simplify inventory control

Not all products are available for sale in all territories. Please inquire.

VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand.

For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
0.5 McFarland StandardH2O (Part No. T3339)
Antibiotic CodeVarious
Antimicrobial AgentsVarious
Broth TypeMHB (Part No. T3462)
DescriptionSensititre Gram Positive All-in-One Plate
Incubation Conditions35°C O2
Incubation Time18-24 hr
Isolates Per Plate1
No. per Pack10
Quantity10/Pk.
Reading OptionsAutoread or manual
Shelf Life18 to 24 Months
Transfer Volume10µL or 30µL* (*Aid in detection of resistance mechanisms for gram positive organisms; see pack insert)
Volume Per Well50 μL
Product TypeAST Plate
Target Organism ClassNon-fastidious, Gram-negative Organisms
Test TypeMinimum Inhibitory Concentration (MIC)
Unit SizeEach