Ion AmpliSeq™ Polyploidy Panel Kit with Ion 520™ Chips (80 Samples)
Ion Torrent™

Ion AmpliSeq™ Polyploidy Panel Kit with Ion 520™ Chips (80 Samples)

The Ion GeneStudio System–compatible Ion AmpliSeq Polyploidy Panel Kit empowers accurate quality controls in preimplantation genetic testing for aneuploidy (PGT-A)Read more
Have Questions?
Catalog NumberQuantity
GS4569180 Samples
Catalog number GS45691
Price (USD)
5,320.00
Each
Quantity:
80 Samples
Price (USD)
5,320.00
Each

The Ion GeneStudio System–compatible Ion AmpliSeq Polyploidy Panel Kit empowers accurate quality controls in preimplantation genetic testing for aneuploidy (PGT-A) by leveraging SNP-based assessments.

Benefits of the Ion AmpliSeq Polyploidy Panel Kit include:
• Germline SNP detection from embryo biopsy samples—no parental DNA required, enabling the extraction of genetic insights directly from the sample
• Comprehensive genetic coverage—comprises 74 microhaplotype amplicons for human identification and 368 single SNP amplicons representing the population at large. With a total of 590 SNP sites covered, the panel unveils the unique genomic variations present in each sample, facilitating premium PGT-A analysis.
• Triploidy 69,XXX identification—through the utilization of tri-allelic microhaplotype sites and allele frequency distributions, samples with the potentially lethal triploidy 69,XXX can be confidently identified. This abnormality often appears similar to the normal 46,XX on conventional PGT-A plots, but the panel's advanced techniques help overcome this challenge.
• Mitigation of maternal contamination risk—reliable assessment of contamination from cumulus cell DNA made possible by tri-allelic microhaplotype sites. This assessment assists in prioritizing low-risk samples with minimized maternal contamination concerns.
• Sibling embryo tracking—SNP clustering allows for the identification of genetically related embryos, helping prevent sample mix-ups.

Enhance embryo prioritization in PGT-A through the implementation of SNP-based quality controls
Single nucleotide polymorphisms (SNPs) are unique genetic variations that can be inherited and directly associated with parental DNA. By leveraging SNPs, PGT-A analysis gains valuable insights into triploidy 69,XXX, maternal contamination, helping ease the selection of embryos for IVF research.

Notes
• This kit is compatible with the Ion GeneStudio System.
 

For Research Use Only.
Specifications
Quantity80 Samples
FormatKit
Sequencing TypeNext Generation Sequencing
Unit SizeEach
Contents & Storage
• Ion AmpliSeq Polyploidy Panel, -10°C to -30°C
• Ion AmpliSeq Lib Kit Plus, -10°C to -30°C
• Ion S5 Chef Supplies, 15°C to 30°C
• Ion S5 Chef Solutions, 15°C to 30°C
• Ion S5 Chef Reagents, -10°C to -30°C
• Ion S5 Sequencing Solutions, 15°C to 30°C
• Ion S5 Sequencing Reagents, -10°C to -30°C
• Ion 520 Chip Kit, 15°C to 30°C

Frequently asked questions (FAQs)

What is the turnaround time (TAT) for running 48 samples on the Ion GX5 chip?

The turnaround time (TAT) for running 48 samples on the Ion GX5 chip is 18 hours.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

Using the Ion AmpliSeq Polyploidy Panel Kit with Ion GX5 Chips (384 Samples) (Cat. No. GX55689), can I run samples from Ion ReproSeq PGT-A library and Ion AmpliSeq Polyploidy Panel library in the same lane of the Ion GX5 chip?

Samples from Ion ReproSeq PGT-A and Ion AmpliSeq Polyploidy Panel libraries must be run in separate lanes of the Ion GX5 chip.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

How many samples in total can I run with the Ion AmpliSeq Polyploidy Panel Kit with Ion GX5 Chips (384 Samples) (Cat. No. GX55689)?

The kit is sufficient for running up to 384 samples since 2 chips are included in the kit with each chip having 4 lanes where one can run up to 48 samples per lane.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

How many samples in total can I run with Ion AmpliSeq Polyploidy Panel Kit with Ion 520 Chips (80 Samples) (Cat. No. GS45691)?

The kit is sufficient for running up to 80 samples since 4 chips are included in the kit and each chip can run up to 20 samples.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

How should I store the Ion AmpliSeq Polyploidy Panel?

Ion AmpliSeq Polyploidy Panel should be stored at -10 to -30 degrees C.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.