Owl™ Gel Staining Box
Thermo Scientific™

Owl™ Gel Staining Box

Stain, fix and de-stain in the same container with Thermo Scientific™ Owl™ Gel Staining Boxes, offering a convenient, conventional staining method.
Have Questions?
Catalog NumberQuantity
GSB-3Pk. of 5 boxes
Catalog number GSB-3
Price (USD)
77.63
Online exclusive
87.10
Save 9.47
Each
Quantity:
Pk. of 5 boxes
Price (USD)
77.63
Online exclusive
87.10
Save 9.47
Each

Stain, fix and de-stain in the same container with Thermo Scientific™ Owl™ Gel Staining Boxes, offering a convenient, conventional staining method. Boxes hold up to 125mL of liquid and one gel when used on a shaking platform.

  • Trays available in convenient size to hold the proper volume of staining reagents
  • Grooved lid enables proper orientation and stability when stacking
  • Made of clear plastic, so gels can be viewed directly in the tray

Ordering Information:

Package of five

Specifications
DescriptionGel Staining Box; Pack of 5
Height (Metric)3 cm
Length (Metric)11 cm
QuantityPk. of 5 boxes
Volume (Metric)125 mL
Width (Metric)11 cm
Unit SizeEach