Easily identify cell nuclei and other features of interest during high-content screening (HCS) assays with the versatile HCS NuclearMask Blue, Red, and Deep Red stains, which can be used with live and fixed cells and in multiparametric analyses.
Have Questions?
Change view
Catalog Number
Color
H10326
Red
H10325
Blue
H10294
Far-Red, Deep Red
3 Options
Catalog number H10326
Price (USD)
362.65
Online exclusive
374.00
Save 11.35
Each
Color:
Red
Price (USD)
362.65
Online exclusive
374.00
Save 11.35
Each
Easily identify cell nuclei in high-content screening (HCS) assays with the versatile HCS NuclearMask Blue, Red, and Deep Red stains, which survive standard formaldehyde-based fixation and detergent-based permeabilization methods and can be applied to live and fixed cells. Because their dyes are easily discriminated from other fluorophores, the HCS NuclearMask stains can be used in multiparametric analyses. They can also label the entire cell (i.e., cytoplasm and nucleus) and provide an accurate backdrop against which to assess the features of interest.
The versatile HCS NuclearMask Blue, HCS NuclearMask Red, and HCS NuclearMask Deep Red stains can be used to measure DNA content and perform cell demarcation with formaldehyde-fixed cells on high content imaging and analysis (HCS) platforms. HCS NuclearMask Blue, HCS NuclearMask Red, and HCS NuclarMask Deep Red stains have excitation/emission maxima of 355/460 nm, 622/645 nm, and 638/686 nm, respectively, when bound to DNA. HCS NuclearMask Blue stain is easily discriminated from visible- and near-IR wavelength fluorophores, HCS NuclearMask Red stain is easily discriminated from green- and even orange-fluorescent fluorophores, and HCS NuclearMask Deep Red is easily discriminated from visible wavelength fluorophores when used in multiparametric analyses.
If total cell staining is desired, HCS CellMask stains label the entire cell (cytoplasm and nucleus) and provide an accurate backdrop against which the features of interest can be assessed. HCS CellMask stains are available in a choice of colors for multiplexing flexibility. Sufficient volume quantities are provided to stain ten 96-well plates using an assay volume of 100 μL per well.
For Research Use Only. Not for human or animal therapeutic or diagnostic use.
Specifications
ColorRed
DescriptionHCS NuclearMask™ Red Stain
Detection MethodFluorescence
EmissionVisible
Excitation Wavelength Range622/645
For Use With (Equipment)High Content Instrument
Product LineNuclearMask
Quantity125 μL
Shipping ConditionRoom Temperature
Label TypeFluorescent Dye
Product TypeStain
Sub Cellular LocalizationNucleic Acids, Nucleus
Unit SizeEach
Contents & Storage
Includes 125 μL of 1000X concentrate, providing sufficient material to stain a total of ten 96-well microplates.
Store at ≤-20°C, desiccate, and protect from light.
Citations & References (5)
Citations & References
Abstract
CD40L induces functional tunneling nanotube networks exclusively in dendritic cells programmed by mediators of type 1 immunity.
The ability of dendritic cells (DC) to mediate CD4(+) T cell help for cellular immunity is guided by instructive signals received during DC maturation, as well as the resulting pattern of DC responsiveness to the Th signal, CD40L. Furthermore, the professional transfer of antigenic information from migratory DC to lymph ... More
Overcoming Adaptive Resistance to KRAS and MEK Inhibitors by Co-targeting mTORC1/2 Complexes in Pancreatic Cancer.
Authors:
Journal:Cell Rep Med
PubMed ID:33294856
Genome-wide synthetic lethal screen unveils novel CAIX-NFS1/xCT axis as a targetable vulnerability in hypoxic solid tumors.
Authors:
Journal:Sci Adv
PubMed ID:34452919
cAMP and EPAC Signaling Functionally Replace OCT4 During Induced Pluripotent Stem Cell Reprogramming.
Authors:
Journal:Mol Ther
PubMed ID:25666918
A medium-throughput analysis of signaling pathways involved in early stages of stem cell reprogramming.