Can Image-iT Red Hypoxia Reagent be used in cells expressing fluorescent proteins?
Yes, as long as the fluorescent protein does not emit in the red range (610 nm). For cells that do not express a fluorescent protein, we recommend viewing unstained cells under the TRITC and Texas Red channels to examine autofluorescence. For cells expressing a fluorescent protein, we recommend analyzing unstained cell samples to determine the extent the fluorescent protein emission may overlap in these channels.