Ethyl 5-phenoxycarbonylamino-1,2,3-thiadiazole-4-carboxylate, 98+%
Ethyl 5-phenoxycarbonylamino-1,2,3-thiadiazole-4-carboxylate, 98+%
Ethyl 5-phenoxycarbonylamino-1,2,3-thiadiazole-4-carboxylate, 98+%
Ethyl 5-phenoxycarbonylamino-1,2,3-thiadiazole-4-carboxylate, 98+%
Thermo Scientific Chemicals

Ethyl 5-phenoxycarbonylamino-1,2,3-thiadiazole-4-carboxylate, 98+%

CAS: 2037-81-2 | C12H11N3O4S | 293.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H25763.03
also known as H25763-03
Price (USD)
154.65
Online Exclusive
172.00
Save 17.35 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
154.65
Online Exclusive
172.00
Save 17.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS2037-81-2
IUPAC Nameethyl 5-[(phenoxycarbonyl)amino]-1,2,3-thiadiazole-4-carboxylate
Molecular FormulaC12H11N3O4S
InChI KeyFCPHNPRVMWSGSK-UHFFFAOYSA-N
SMILESCCOC(=O)C1=C(NC(=O)OC2=CC=CC=C2)SN=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥98.0%
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Melting Point (clear melt)150.5-156.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only