2,4-Dimethoxypyrimidine, 98+%
2,4-Dimethoxypyrimidine, 98+%
2,4-Dimethoxypyrimidine, 98+%
Thermo Scientific Chemicals

2,4-Dimethoxypyrimidine, 98+%

CAS: 3551-55-1 | C6H8N2O2 | 140.142 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H25799.03
also known as H25799-03
Price (USD)
28.80
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
28.80
Each
Chemical Identifiers
CAS3551-55-1
IUPAC Name2,4-dimethoxypyrimidine
Molecular FormulaC6H8N2O2
InChI KeyKEVRHVMWBKFGLO-UHFFFAOYSA-N
SMILESCOC1=CC=NC(OC)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥98.0%
Refractive Index1.5005-1.5045 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only