2,2-Difluorobutyric acid, 97%
2,2-Difluorobutyric acid, 97%
2,2-Difluorobutyric acid, 97%
2,2-Difluorobutyric acid, 97%
Thermo Scientific Chemicals

2,2-Difluorobutyric acid, 97%

CAS: 2366-62-3 | C4H6F2O2 | 124.087 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H25830.MD
also known as H25830-MD
Price (USD)
50.50
Each
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)
50.50
Each
Add to cart
Chemical Identifiers
CAS2366-62-3
IUPAC Name2,2-difluorobutanoic acid
Molecular FormulaC4H6F2O2
InChI KeySBSKRVZFWQPJID-UHFFFAOYSA-N
SMILESCCC(F)(F)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Refractive Index1.3615-1.3665 @ 20?C
Appearance (Color)Clear colorless
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only