3-Methylthio-1-propylamine, 97%
3-Methylthio-1-propylamine, 97%
3-Methylthio-1-propylamine, 97%
3-Methylthio-1-propylamine, 97%
Thermo Scientific Chemicals

3-Methylthio-1-propylamine, 97%

CAS: 4104-45-4 | C4H11NS | 105.199 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H25846.06
also known as H25846-06
Price (USD)
144.65
Online Exclusive
161.00
Save 16.35 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
144.65
Online Exclusive
161.00
Save 16.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS4104-45-4
IUPAC Name3-(methylsulfanyl)propan-1-aminium
Molecular FormulaC4H12NS
InChI KeyKKYSBGWCYXYOHA-UHFFFAOYSA-O
SMILESCSCCC[NH3+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Assay (GC)≥96.0%
Refractive Index1.4870-1.4940 @ 20°C
FormLiquid
3-Aminopropyl Methyl Sulfideis used in cheese and seafood flavors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Aminopropyl Methyl Sulfideis used in cheese and seafood flavors.

Solubility
Insoluble in water. Soluble in ethanol.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
RUO – Research Use Only
Kullberg, D.W. ; Orr, G.R. ; Coward, J.K. Stereochemical course of enzyme-catalyzed aminopropyl transfer: spermidine synthase Fed. Proc., Fed. Am. Soc. Exp. Biol.. 1986, 45