(+/-)-2-(Trifluoromethyl)pyrrolidine, 95%
(+/-)-2-(Trifluoromethyl)pyrrolidine, 95%
(+/-)-2-(Trifluoromethyl)pyrrolidine, 95%
Thermo Scientific Chemicals

(+/-)-2-(Trifluoromethyl)pyrrolidine, 95%

CAS: 109074-67-1 | C5H8F3N | 139.121 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H25852.03
also known as H25852-03
Price (USD)
220.65
Special offer
Online exclusive
Ends: 30-Jun-2026
259.00
Save 38.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
220.65
Special offer
Online exclusive
Ends: 30-Jun-2026
259.00
Save 38.35 (15%)
Each
Chemical Identifiers
CAS109074-67-1
IUPAC Name2-(trifluoromethyl)pyrrolidine
Molecular FormulaC5H8F3N
InChI KeyVINAMCOZNJHNIH-UHFFFAOYNA-N
SMILESFC(F)(F)C1CCCN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥94.0%
Refractive Index1.3760-1.3820 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only