(E)-3,4-Dihydroxybenzylideneacetone, 97%
(E)-3,4-Dihydroxybenzylideneacetone, 97%
(E)-3,4-Dihydroxybenzylideneacetone, 97%
Thermo Scientific Chemicals

(E)-3,4-Dihydroxybenzylideneacetone, 97%

CAS: 123694-03-1 | C10H10O3 | 178.187 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H25901.03
also known as H25901-03
Price (USD)
77.65
Special offer
Online exclusive
Ends: 30-Jun-2026
91.00
Save 13.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
77.65
Special offer
Online exclusive
Ends: 30-Jun-2026
91.00
Save 13.35 (15%)
Each
Chemical Identifiers
CAS123694-03-1
IUPAC Name4-(3,4-dihydroxyphenyl)but-3-en-2-one
Molecular FormulaC10H10O3
InChI KeyYIFZKRGUGKLILR-UHFFFAOYSA-N
SMILESCC(=O)C=CC1=CC(O)=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to pale brown
Formpowder
Assay (GC)> 96.0%
Purity (DSC)> 96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only