Ethyl 2-ethoxymethylene-4,4,4-trifluoro-3-oxobutyrate, 97%
Ethyl 2-ethoxymethylene-4,4,4-trifluoro-3-oxobutyrate, 97%
Ethyl 2-ethoxymethylene-4,4,4-trifluoro-3-oxobutyrate, 97%
Thermo Scientific Chemicals

Ethyl 2-ethoxymethylene-4,4,4-trifluoro-3-oxobutyrate, 97%

CAS: 571-55-1 | C9H11F3O4 | 240.178 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number H25927.09
also known as H25927-09
Price (USD)
118.65
Special offer
Online exclusive
Ends: 28-Dec-2026
139.00
Save 20.35
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
118.65
Special offer
Online exclusive
Ends: 28-Dec-2026
139.00
Save 20.35
Each
Chemical Identifiers
CAS571-55-1
IUPAC Nameethyl 2-(ethoxymethylidene)-4,4,4-trifluoro-3-oxobutanoate
Molecular FormulaC9H11F3O4
InChI KeyXNGGOXOLHQANRB-UHFFFAOYSA-N
SMILESCCOC=C(C(=O)OCC)C(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4270-1.4320 @ 20?C
FormLiquid
Assay (GC)≥96.0%
Appearance (Color)Clear colorless to pale yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only