meso-Tetrakis(4-methoxyphenyl)porphine, 95%
meso-Tetrakis(4-methoxyphenyl)porphine, 95%
meso-Tetrakis(4-methoxyphenyl)porphine, 95%
Thermo Scientific Chemicals

meso-Tetrakis(4-methoxyphenyl)porphine, 95%

CAS: 22112-78-3 | C48H38N4O4 | 734.856 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Catalog number H25946.09
also known as H25946-09
Price (USD)
283.65
Special offer
Online exclusive
Ends: 30-Jun-2026
334.00
Save 50.35 (15%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
283.65
Special offer
Online exclusive
Ends: 30-Jun-2026
334.00
Save 50.35 (15%)
Each
Chemical Identifiers
CAS22112-78-3
IUPAC Name2,7,12,17-tetrakis(4-methoxyphenyl)-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1,3,5,7,9,11,13(22),14,16,18(21),19-undecaene
Molecular FormulaC48H38N4O4
InChI KeySKXWDGDNWBYACJ-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1)C1=C2NC(C=C2)=C(C2=NC(C=C2)=C(C2=NC(C=C2)=C(C2=CC=C1N2)C1=CC=C(OC)C=C1)C1=CC=C(OC)C=C1)C1=CC=C(OC)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Purple to dark purple
FormPowder
Assay (unspecified)(Based on C, H & N Elemental Analysis): >94.0 to <106.0%
Elemental AnalysisCarbon C: 73.7-83.1%
lambda-max (UV-Vis)420-424nm (0.1% in chloroform)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only