2-Aminoimidazole sulfate, 98%
2-Aminoimidazole sulfate, 98%
2-Aminoimidazole sulfate, 98%
Thermo Scientific Chemicals

2-Aminoimidazole sulfate, 98%

CAS: 1450-93-7 | C6H12N6O4S | 264.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Catalog number H25999.09
also known as H25999-09
Price (USD)
271.65
Online Exclusive
302.00
Save 30.35 (10%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
271.65
Online Exclusive
302.00
Save 30.35 (10%)
Each
Chemical Identifiers
CAS1450-93-7
IUPAC Namebis(1H-imidazol-2-amine); sulfuric acid
Molecular FormulaC6H12N6O4S
InChI KeyKUWRLKJYNASPQZ-UHFFFAOYSA-N
SMILESOS(O)(=O)=O.NC1=NC=CN1.NC1=NC=CN1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
Identification (FTIR)Conforms
Appearance (Color)White to pale cream to pale brown
FormCrystals or powder or crystalline powder
Assay (HPLC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only