2-Amino-5-phenylpyridine, 97%
2-Amino-5-phenylpyridine, 97%
2-Amino-5-phenylpyridine, 97%
Thermo Scientific Chemicals

2-Amino-5-phenylpyridine, 97%

CAS: 33421-40-8 | C11H10N2 | 170.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H26002.MD
also known as H26002-MD
Price (USD)
54.65
Special offer
Online exclusive
Ends: 30-Jun-2026
64.60
Save 9.95 (15%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
54.65
Special offer
Online exclusive
Ends: 30-Jun-2026
64.60
Save 9.95 (15%)
Each
Chemical Identifiers
CAS33421-40-8
IUPAC Name5-phenylpyridin-2-amine
Molecular FormulaC11H10N2
InChI KeyOAPVIBHQRYFYSE-UHFFFAOYSA-N
SMILESNC1=CC=C(C=N1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to pale yellow
FormCrystals or powder or crystalline powder
Melting Point (clear melt)131.0-140.0?C
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only