4-(tert-Butoxy)phenol, 98%
4-(tert-Butoxy)phenol, 98%
4-(tert-Butoxy)phenol, 98%
4-(tert-Butoxy)phenol, 98%
Thermo Scientific Chemicals

4-(tert-Butoxy)phenol, 98%

CAS: 2460-87-9 | C10H14O2 | 166.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H26013.06
also known as H26013-06
Price (USD)
274.65
Special offer
Online exclusive
Ends: 30-Jun-2026
323.00
Save 48.35 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
274.65
Special offer
Online exclusive
Ends: 30-Jun-2026
323.00
Save 48.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS2460-87-9
IUPAC Name4-(tert-butoxy)phenol
Molecular FormulaC10H14O2
InChI KeyCIICLJLSRUHUBY-UHFFFAOYSA-N
SMILESCC(C)(C)OC1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale brown to gray
Assay (GC)≥97.5%
Melting Point (clear melt)149.0-156.0?C
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only