4-Chloro-alpha-toluenesulfonyl chloride, 97%
4-Chloro-alpha-toluenesulfonyl chloride, 97%
4-Chloro-alpha-toluenesulfonyl chloride, 97%
Thermo Scientific Chemicals

4-Chloro-alpha-toluenesulfonyl chloride, 97%

CAS: 6966-45-6 | C7H6Cl2O2S | 225.083 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H26017.03
also known as H26017-03
Price (USD)
63.65
Special offer
Online exclusive
Ends: 30-Jun-2026
75.20
Save 11.55 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
63.65
Special offer
Online exclusive
Ends: 30-Jun-2026
75.20
Save 11.55 (15%)
Each
Chemical Identifiers
CAS6966-45-6
IUPAC Name(4-chlorophenyl)methanesulfonyl chloride
Molecular FormulaC7H6Cl2O2S
InChI KeyDBJRPJSDYFDWPV-UHFFFAOYSA-N
SMILESClC1=CC=C(CS(Cl)(=O)=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
Melting Point (clear melt)90.0-99.0?C
Appearance (Color)White to pale cream to pale yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only