alpha,alpha-Difluorophenylacetic acid, 97%
alpha,alpha-Difluorophenylacetic acid, 97%
alpha,alpha-Difluorophenylacetic acid, 97%
Thermo Scientific Chemicals

alpha,alpha-Difluorophenylacetic acid, 97%

CAS: 360-03-2 | C8H6F2O2 | 172.131 g/mol
Have Questions?
Catalog NumberQuantity
H26028.MD
also known as H26028-MD
250 mg
Catalog number H26028.MD
also known as H26028-MD
Price (USD)
54.65
Special offer
Online exclusive
Ends: 30-Jun-2026
63.90
Save 9.25 (14%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
54.65
Special offer
Online exclusive
Ends: 30-Jun-2026
63.90
Save 9.25 (14%)
Each
Chemical Identifiers
CAS360-03-2
IUPAC Name2-(2,3-difluorophenyl)acetate
Molecular FormulaC8H5F2O2
InChI KeyUXSQXUSJGPVOKT-UHFFFAOYSA-M
SMILES[O-]C(=O)CC1=CC=CC(F)=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)71-77?C
Appearance (Color)White to cream or pale yellow
FormCrystals or crystalline powder
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Assay (Silylated GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only