4'-Fluoroisobutyrophenone, 97%
4'-Fluoroisobutyrophenone, 97%
4'-Fluoroisobutyrophenone, 97%
4'-Fluoroisobutyrophenone, 97%
Thermo Scientific Chemicals

4'-Fluoroisobutyrophenone, 97%

CAS: 26393-91-9 | C10H11FO | 166.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H26036.03
also known as H26036-03
Price (USD)
153.65
Online Exclusive
170.00
Save 16.35 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
153.65
Online Exclusive
170.00
Save 16.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS26393-91-9
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Refractive Index1.4960-1.5010 @ 20?C
FormLiquid
Appearance (Color)Clear colorless to pale yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only