2-(1,3-Dioxolan-2-yl)ethylboronic acid pinacol ester, 97%
2-(1,3-Dioxolan-2-yl)ethylboronic acid pinacol ester, 97%
2-(1,3-Dioxolan-2-yl)ethylboronic acid pinacol ester, 97%
Thermo Scientific Chemicals

2-(1,3-Dioxolan-2-yl)ethylboronic acid pinacol ester, 97%

CAS: 1073354-07-0 | C11H21BO4 | 228.095 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H26071.06
also known as H26071-06
Price (USD)
238.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
238.00
Each
Specifications
CAS1073354-07-0
Chemical Name or Material2-(1,3-Dioxolan-2-yl)ethylboronic acid pinacol ester
Formula Weight228.1
IUPAC Name2-[2-(1,3-dioxolan-2-yl)ethyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
InChI KeyDNBRLKJRBDIKOO-UHFFFAOYSA-N
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only