3-Pyrroline, tech. 85% (remainder pyrrolidine)
3-Pyrroline, tech. 85% (remainder pyrrolidine)
3-Pyrroline, tech. 85% (remainder pyrrolidine)
Thermo Scientific Chemicals

3-Pyrroline, tech. 85% (remainder pyrrolidine)

CAS: 109-96-6 | C4H7N | 69.107 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H26072.03
also known as H26072-03
Price (USD)
43.65
Special offer
Online exclusive
Ends: 28-Dec-2026
51.20
Save 7.55
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
43.65
Special offer
Online exclusive
Ends: 28-Dec-2026
51.20
Save 7.55
Each
Chemical Identifiers
CAS109-96-6
IUPAC Name2,5-dihydro-1H-pyrrole
Molecular FormulaC4H7N
InChI KeyJVQIKJMSUIMUDI-UHFFFAOYSA-N
SMILESC1NCC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Water Content (Karl Fischer Titration)≤2.0%
Impurity contentPyrrolidine <15% by ¹H-NMR, typically 10-13%
Refractive Index1.4650-1.4730 @ 20°C
Appearance (Color)Clear colorless to pale yellow
Assay (unspecified)≥83.0% with pyrrolidine
View more
3-Pyrroline is suitable to the core-level binding energies of simple unsaturated organic molecules bonded to the Si(001) surface by X-ray photoelectron spectroscopy (XPS). It is used in the synthesis of renin inhibitors and vasodilators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Pyrroline is suitable to the core-level binding energies of simple unsaturated organic molecules bonded to the Si(001) surface by X-ray photoelectron spectroscopy (XPS). It is used in the synthesis of renin inhibitors and vasodilators.

Solubility
Soluble in alcohol and ether. Miscible with water.

Notes
Air sensitive. Store at -20°C. Store away from oxidizing agents, air.
RUO – Research Use Only

General References:

  1. Martin P. Green; Jeremy C. Prodger; Alexandra E. Sherlock and Christopher J. Hayes. A Convenient Method for 3-Pyrroline Synthesis. Org. Lett. 2001, 3 (21), 1-4.
  2. Masahiro Sai and Seijiro Matsubara. Silver-Catalyzed Intramolecular Chloroamination of Allenes: Easy Access to Functionalized 3-Pyrroline and Pyrrole Derivatives. Org. Lett. 2011, 13 (17), 4676-4679.