Ethyl 4-cyanophenylacetate, 98%, Thermo Scientific Chemicals
Ethyl 4-cyanophenylacetate, 98%, Thermo Scientific Chemicals
Ethyl 4-cyanophenylacetate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 4-cyanophenylacetate, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H26078.03
also known as H26078-03
1 g
Catalog number H26078.03
also known as H26078-03
Price (USD)
94.65
Special offer
Online exclusive
Ends: 28-Dec-2026
111.00
Save 16.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
94.65
Special offer
Online exclusive
Ends: 28-Dec-2026
111.00
Save 16.35
Each
Chemical Identifiers
CAS1528-41-2
IUPAC Nameethyl 2-(4-cyanophenyl)acetate
Molecular FormulaC11H11NO2
InChI KeyDFEWKWBIPMKGFG-UHFFFAOYSA-N
SMILESCCOC(=O)CC1=CC=C(C=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)White to yellow to green to orange
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only