4-Fluoro-2-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals
4-Fluoro-2-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals
4-Fluoro-2-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Fluoro-2-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H26091.03
also known as H26091-03
1 g
Catalog number H26091.03
also known as H26091-03
Price (USD)
85.65
Online Exclusive
95.00
Save 9.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
85.65
Online Exclusive
95.00
Save 9.35 (10%)
Each
Chemical Identifiers
CAS450-83-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder
Free acid (titration)≤1.0%
Melting Point (clear melt)58.5-64.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only