2,6-Difluoro-4-hydroxybenzoic acid, 95%
2,6-Difluoro-4-hydroxybenzoic acid, 95%
2,6-Difluoro-4-hydroxybenzoic acid, 95%
Thermo Scientific Chemicals

2,6-Difluoro-4-hydroxybenzoic acid, 95%

CAS: 214917-68-7 | C7H4F2O3 | 174.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H26134.03
also known as H26134-03
Price (USD)
132.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
132.00
Each
Chemical Identifiers
CAS214917-68-7
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (Silylated GC)≥94.0%
Appearance (Color)White to cream
Assay (Aqueous acid-base Titration)≥94.0 to ≤106.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only