2-Fluoro-4-hydroxybenzaldehyde, 97%
2-Fluoro-4-hydroxybenzaldehyde, 97%
2-Fluoro-4-hydroxybenzaldehyde, 97%
2-Fluoro-4-hydroxybenzaldehyde, 97%
Thermo Scientific Chemicals

2-Fluoro-4-hydroxybenzaldehyde, 97%

CAS: 348-27-6 | C7H5FO2 | 140.113 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
Catalog number H26218.ME
also known as H26218-ME
Price (USD)
303.65
Online Exclusive
337.00
Save 33.35 (10%)
Each
Add to cart
Quantity:
500 mg
Request bulk or custom format
Price (USD)
303.65
Online Exclusive
337.00
Save 33.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS348-27-6
IUPAC Name2-fluoro-4-hydroxybenzaldehyde
Molecular FormulaC7H5FO2
InChI KeyONRPXRPUBXXCCM-UHFFFAOYSA-N
SMILESOC1=CC(F)=C(C=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)Cream or pink to gray
Assay (GC)≥96.0%
Melting Point (clear melt)165.0-175.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only