4-(Methylthio)phenol, 98%
4-(Methylthio)phenol, 98%
4-(Methylthio)phenol, 98%
Thermo Scientific Chemicals

4-(Methylthio)phenol, 98%

CAS: 1073-72-9 | C7H8OS | 140.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number H26225.09
also known as H26225-09
Price (USD)
49.65
Online Exclusive
54.80
Save 5.15 (9%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
49.65
Online Exclusive
54.80
Save 5.15 (9%)
Each
Chemical Identifiers
CAS1073-72-9
IUPAC Name4-(methylsulfanyl)phenol
Molecular FormulaC7H8OS
InChI KeyQASBCTGZKABPKX-UHFFFAOYSA-N
SMILESCSC1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)White to pale brown
FormCrystals or powder or crystalline powder or flakes
Assay (GC)≥97.5%
4-(Methylthio)phenol is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Methylthio)phenol is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in water 9.59 g/ l.

Notes
Store in a cool, dry place in tightly closed container. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Brian John. Johnson; Therese A. N-Acylamino acid esters of 4-(methylthio)phenol. J. Org. Chem. 1970, 35 (1), 255-257.
  2. Brian J. Johnson; Elwood G. Trask. 4-(Methylthio)phenyl and 4-(methylsulfonyl)phenyl esters in the preparation of peptides and polypeptides. II. Synthesis of the protected heptapeptide (A82-A88) of bovine chymotrypsinogen A. J. Org. Chem. 1968, 33 (12), 4521-4524.