1-Bromo-4-n-propylbenzene, 99%
1-Bromo-4-n-propylbenzene, 99%
1-Bromo-4-n-propylbenzene, 99%
Thermo Scientific Chemicals

1-Bromo-4-n-propylbenzene, 99%

CAS: 588-93-2 | C9H11Br | 199.091 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number H26236.14
also known as H26236-14
Price (USD)
107.65
Online Exclusive
119.00
Save 11.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
107.65
Online Exclusive
119.00
Save 11.35 (10%)
Each
Chemical Identifiers
CAS588-93-2
IUPAC Name1-bromo-4-propylbenzene
Molecular FormulaC9H11Br
InChI KeyNUPWGLKBGVNSJX-UHFFFAOYSA-N
SMILESCCCC1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5345-1.5375 @ 20?C
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only