Thermo Scientific Chemicals

Dimethyl cyclohexylmalonate, 95%, Thermo Scientific Chemicals

Catalog number: H26289.03
1 g, Each
Thermo Scientific Chemicals

Dimethyl cyclohexylmalonate, 95%, Thermo Scientific Chemicals

Catalog number: H26289.03
1 g, Each
Quantity
Catalog number: H26289.03
also known as H26289-03
Price (USD)
Price: 88.80
Online price: 76.65
Your price:
Quantity
-

Chemical Identifiers

CAS
49769-76-8
IUPAC Name
1,3-dimethyl 2-cyclohexylpropanedioate
Molecular Formula
C11H18O4
InChI Key
CULITIDIVVTWIV-UHFFFAOYSA-N
SMILES
COC(=O)C(C1CCCCC1)C(=O)OC
Form
Liquid
Refractive Index
1.4540-1.4600 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥94.0%

Description

Used in developing a one-pot, two-step procedure for decarbalkoxylation of malonic esters. The effect of 15C5, other cations, and good nucleophiles was also studied.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in developing a one-pot, two-step procedure for decarbalkoxylation of malonic esters. The effect of 15C5, other cations, and good nucleophiles was also studied.

Solubility
Water soluble.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text