Dimethyl cyclohexylmalonate, 95%
Dimethyl cyclohexylmalonate, 95%
Dimethyl cyclohexylmalonate, 95%
Thermo Scientific Chemicals

Dimethyl cyclohexylmalonate, 95%

CAS: 49769-76-8 | C11H18O4 | 214.261 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H26289.06
also known as H26289-06
Price (USD)
259.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
259.00
Each
Chemical Identifiers
CAS49769-76-8
IUPAC Name1,3-dimethyl 2-cyclohexylpropanedioate
Molecular FormulaC11H18O4
InChI KeyCULITIDIVVTWIV-UHFFFAOYSA-N
SMILESCOC(=O)C(C1CCCCC1)C(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4540-1.4600 @ 20?C
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥94.0%
Used in developing a one-pot, two-step procedure for decarbalkoxylation of malonic esters. The effect of 15C5, other cations, and good nucleophiles was also studied.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in developing a one-pot, two-step procedure for decarbalkoxylation of malonic esters. The effect of 15C5, other cations, and good nucleophiles was also studied.

Solubility
Water soluble.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. A. Paul Krapcho. Recent synthetic applications of the dealkoxycarbonylation reaction. Part 1. Dealkoxycarbonylations of malonate esters. ARKIVOC. 2007, 2 1-53.
  2. Duncan H. Hunter,; Vijay Patel,; Richard A. Perry. Crown ether catalysis of decarboxylation and decarbalkoxylation of β-keto acids and malonates: a synthetic application. Canadian Journal of Chemistry. 1980, 58 (21),2271-2277.