4-Bromobenzyl mercaptan, 98%
4-Bromobenzyl mercaptan, 98%
4-Bromobenzyl mercaptan, 98%
Thermo Scientific Chemicals

4-Bromobenzyl mercaptan, 98%

CAS: 19552-10-4 | C7H7BrS | 203.097 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number H26299.03
also known as H26299-03
Price (USD)
102.65
Special offer
Online exclusive
Ends: 28-Dec-2026
121.00
Save 18.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
102.65
Special offer
Online exclusive
Ends: 28-Dec-2026
121.00
Save 18.35
Each
Chemical Identifiers
CAS19552-10-4
IUPAC Name(4-bromophenyl)methanethiol
Molecular FormulaC7H7BrS
InChI KeyCUCKXDPCCYHFMQ-UHFFFAOYSA-N
SMILESSCC1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormFused low melting solid (Clear to turbid liquid as melt)
Appearance (Color)White to cream or pale yellow
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only