Ethyl propenyl ether, cis + trans, 97%
Ethyl propenyl ether, cis + trans, 97%
Ethyl propenyl ether, cis + trans, 97%
Thermo Scientific Chemicals

Ethyl propenyl ether, cis + trans, 97%

CAS: 928-55-2 | C5H10O | 86.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number H26319.14
also known as H26319-14
Price (USD)
91.65
Online Exclusive
102.00
Save 10.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
91.65
Online Exclusive
102.00
Save 10.35 (10%)
Each
Chemical Identifiers
CAS928-55-2
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.3945-1.3995 @ 20?C
Assay (GC)≥96.0%
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only