Diethyl (2,4-dichlorophenyl)malonate, 95%
Diethyl (2,4-dichlorophenyl)malonate, 95%
Diethyl (2,4-dichlorophenyl)malonate, 95%
Thermo Scientific Chemicals

Diethyl (2,4-dichlorophenyl)malonate, 95%

CAS: 111544-93-5 | C13H14Cl2O4 | 305.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
2.5 g
Catalog number H26330.27
also known as H26330-27
Price (USD)
253.65
Online Exclusive
282.00
Save 28.35 (10%)
Each
Quantity:
2.5 g
Request bulk or custom format
Price (USD)
253.65
Online Exclusive
282.00
Save 28.35 (10%)
Each
Chemical Identifiers
CAS111544-93-5
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.5095-1.5155 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only