1-Phenylethyl mercaptan, 98%
1-Phenylethyl mercaptan, 98%
1-Phenylethyl mercaptan, 98%
Thermo Scientific Chemicals

1-Phenylethyl mercaptan, 98%

CAS: 6263-65-6 | C8H10S | 138.228 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Catalog number H26345.09
also known as H26345-09
Price (USD)
231.65
Online Exclusive
257.00
Save 25.35 (10%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
231.65
Online Exclusive
257.00
Save 25.35 (10%)
Each
Chemical Identifiers
CAS6263-65-6
IUPAC Name1-phenylethane-1-thiol
Molecular FormulaC8H10S
InChI KeyQZZBJCFNHPYNKO-UHFFFAOYNA-N
SMILESCC(S)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Identification (FTIR)Conforms
Assay (GC)≥97.5%
Refractive Index1.5560-1.5600 @ 20?C
FormLiquid
The compound is used as a source of the thiol functional group in organic synthesis. It is introduced by S-alkylation to give alkyl benzylthioethers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The compound is used as a source of the thiol functional group in organic synthesis. It is introduced by S-alkylation to give alkyl benzylthioethers.

Solubility
Insoluble in water. Solubility in ethanol.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
RUO – Research Use Only

General References:

  1. E.J. Corey,; Karlene A. Cimprich. Enantioselective routes to chiral benzylic thiols, sulfinic esters and sulfonic acids illustrated by the 1-phenylethyl series. Tetrahedron Letters. 1992, 33 (29), 4099-4102.
  2. László Garamszegia,; Christian Donzela,; Géraldine Carrotb,; Tuan Q. Nguyena,; Jöns Hilbornc. Synthesis of thiol end-functional polystyrene via atom transfer radical polymerization. Reactive and Functional Polymers. 2003, 55 (2),179-183.