2-Bromo-5-(trifluoromethoxy)phenol, 97%
2-Bromo-5-(trifluoromethoxy)phenol, 97%
2-Bromo-5-(trifluoromethoxy)phenol, 97%
2-Bromo-5-(trifluoromethoxy)phenol, 97%
Thermo Scientific Chemicals

2-Bromo-5-(trifluoromethoxy)phenol, 97%

CAS: 205371-26-2 | C7H4BrF3O2 | 257.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H26389.06
also known as H26389-06
Price (USD)
104.65
Online Exclusive
116.00
Save 11.35 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
104.65
Online Exclusive
116.00
Save 11.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS205371-26-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow or pale red
Assay (GC)≥96.0%
Refractive Index1.4790-1.4840 @ 20°C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only