4-Trifluoromethyl-alpha-toluenesulfonyl chloride, 97%, Thermo Scientific Chemicals
4-Trifluoromethyl-alpha-toluenesulfonyl chloride, 97%, Thermo Scientific Chemicals
4-Trifluoromethyl-alpha-toluenesulfonyl chloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Trifluoromethyl-alpha-toluenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H26395.03
also known as H26395-03
1 g
Catalog number H26395.03
also known as H26395-03
Price (USD)
145.65
Special offer
Online exclusive
Ends: 30-Jun-2026
171.00
Save 25.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
145.65
Special offer
Online exclusive
Ends: 30-Jun-2026
171.00
Save 25.35 (15%)
Each
Chemical Identifiers
CAS163295-75-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
Melting Point (clear melt)100.0-110.0?C
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only